Learn More
4-Methoxybenzenesulfonyl chloride, 98+%
Beskrivelse
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methoxybenzenesulfonyl chloride is used as an anti-HIV as Pharmaceutical Intermediates. It also acts as protecting group agent for various nitrogen functions.
Solubility
Soluble in Toluene. Decomposes in water.
Notes
Moisture Sensitive. Store at 4°C. Store in a cool and dark place. Protect from moisture. Store away from strong bases.
Tekniske data
Tekniske data
| Melting Point | 37°C to 42°C |
| Boiling Point | 119°C to 122°C (0.4 mmHg) |
| Flash Point | >110°C (230°F) |
| Quantity | 25 g |
| UN Number | UN3261 |
| Beilstein | 609005 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Soluble in Toluene. Decomposes in water. |
| Formula Weight | 206.65 |
| Percent Purity | ≥98% |
| Vis mere |
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.