missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl carbazate, 97%, Thermo Scientific Chemicals

Product Code. 11320104
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11320104 5 g 5g
11310104 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11320104 Supplier Thermo Scientific Alfa Aesar Supplier No. L00541.06
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 18912-37-3 | Molecular Formula: C9H12N2O3 | Molecular Weight: 196.206 g/mol

It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the derived azide in N-protection and as a starting material for the azide used to introduce the pMZ-amino protection which is removable under mild hydrolytic conditions.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18912-37-3
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.206
MDL Number MFCD00042817
InChI Key JKBMMHKEAGEILO-UHFFFAOYSA-N
Synonym 4-methoxybenzyl carbazate, p-methoxybenzyl carbazate, 4-methoxybenzyl hydrazinecarboxylate, hydrazinecarboxylic acid, 4-methoxyphenyl methyl ester, 4-methoxyphenyl methyl oxycarbohydrazide, 4-methoxyphenyl methoxycarbohydrazide, carbazic acid, p-methoxybenzyl ester, acmc-20amf6, carbazic acid 4-methoxybenzyl ester, 4-methoxybenzyl hydrazinecarboxylate #
PubChem CID 87847
IUPAC Name (4-methoxyphenyl)methyl N-aminocarbamate
SMILES COC1=CC=C(C=C1)COC(=O)NN

Specifications

Melting Point 75°C to 77°C
Quantity 5 g
Beilstein 2052164
Solubility Information Insoluble in water.
Formula Weight 196.21
Percent Purity 97%
Chemical Name or Material 4-Methoxybenzyl carbazate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.