missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl isothiocyanate, 94%

Product Code. 11378745
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378745 1 g 1g
11388745 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378745 Supplier Thermo Scientific Alfa Aesar Supplier No. L12342.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3694-57-3 | C9H9NOS | 179.237 g/mol

4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3694-57-3
Molecular Formula C9H9NOS
Molecular Weight (g/mol) 179.237
MDL Number MFCD00041117
InChI Key IMFQYAJJXFXVMM-UHFFFAOYSA-N
Synonym 4-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxybenzene, 4-methoxybenzylisothiocyanate, benzene, 1-isothiocyanatomethyl-4-methoxy, p-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxy-benzene, chembl81201, 4-methoxybenzyl isothiocyanate 5gr, acmc-1cq6l
PubChem CID 123197
IUPAC Name 1-(isothiocyanatomethyl)-4-methoxybenzene
SMILES COC1=CC=C(C=C1)CN=C=S

Specifications

Density 1.15
Boiling Point 132°C to 133°C (0.7 mmHg)
Odor Irritating
Refractive Index 1.5975
Quantity 1 g
UN Number UN2922
Beilstein 2089549
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 179.24
Percent Purity 94%
Chemical Name or Material 4-Methoxybenzyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.