missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzyl Isothiocyanate, TRC

Product Code. 30464354
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464354 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464354 Supplier Toronto Research Chemicals Supplier No. TRCM226110100MG
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methoxybenzyl Isothiocyanate
CAS 3694-57-3
Molecular Formula C9H9NOS
Quantity 100 mg
Synonym 1-(Isothiocyanatomethyl)-4-methoxy-benzene, Isothiocyanic Acid p-Methoxybenzyl Ester, alpha-Isothiocyanato-p-methoxytoluene
InChI Formula InChI=1S/C9H9NOS/c1-11-9-4-2-8(3-5-9)6-10-7-12/h2-5H,6H2,1H3
SMILES COc1ccc(CN=C=S)cc1
IUPAC Name 1-(isothiocyanatomethyl)-4-methoxybenzene
Molecular Weight (g/mol) 179.24
Formula Weight 179.0405
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.