missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxybenzylmagnesium chloride, 0.25M in 2-MeTHF

Product Code. 15420736
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420736 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15420736 Supplier Thermo Scientific Alfa Aesar Supplier No. H54078.AE
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 38769-92-5 | C8H9ClMgO | 180.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 38769-92-5
Molecular Formula C8H9ClMgO
MDL Number MFCD01311491
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 4-methoxybenzylmagnesium chloride, 4-methoxybenzylmagnesium chloride solution, magnesium, chloro 4-methoxyphenyl methyl, p-methoxybenzylmagnesium chloride, 4-methoxy benzylmagnesium chloride, xhdyucuvuzsism-uhfffaoysa-m, 4-methoxybenzyl magnesium chloride, chloro 4-methoxyphenyl methyl magnesium, 4-methoxybenzylmagnesium chloride 0.25 m in tetrahydrofuran, 4-methoxybenzylmagnesium chloride solution, 0.25 m in thf
InChI Key XHDYUCUVUZSISM-UHFFFAOYSA-M
SMILES COC1=CC=C(C[Mg]Cl)C=C1
IUPAC Name magnesium;1-methanidyl-4-methoxybenzene;chloride
Molecular Weight (g/mol) 180.91
PubChem CID 10965134
Formula Weight 180.92
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 4-Methoxybenzylmagnesium chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.