missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)benzeneboronic acid, 97%

Product Code. 11476570
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476570 1 g 1g
11486570 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476570 Supplier Thermo Scientific Alfa Aesar Supplier No. H27627.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 99768-12-4 | C8H9BO4 | 179.966 g/mol

It is a reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration and cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. It is employed as reagent in Reagent used in preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid, chromenones and their bradykinin B1 antagonistic activity, Pt nanoparticles on Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injection and salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used for tandem-type Pd(II)-catalyzed oxidative Heck reaction and intramolecular C-H amidation sequence, copper-mediated ligandless aerobic fluoroalkylation of arylboronic acids with fluoroalkyl iodides, one-pot ipso-nitration of arylboronic acids, copper-catalyzed nitration and cyclocondensation followed by palladium-phosphine-catalyzed Suzuki-Miyaura coupling. It is employed as reagent in Reagent used in preparation of biaryls via nickel-catalyzed Suzuki-Miyaura cross-coupling reaction of aryl halides with arylboronic acid, chromenones and their bradykinin B1 antagonistic activity, Pt nanoparticles on Photoactive metal-organic frameworks resulting in efficient hydrogen evolution via synergistic photoexcitation and electron injection and salicylate-based thienylbenzoic acids as E. coli methionine aminopeptidase inhibitor.

Solubility
Soluble in methanol.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99768-12-4
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.966
MDL Number MFCD01632203
InChI Key PQCXFUXRTRESBD-UHFFFAOYSA-N
Synonym methyl 4-boronobenzoate, 4-methoxycarbonyl phenylboronic acid, 4-methoxycarbonyl phenyl boronic acid, 4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonylphenyl boronic acid, methyl4-boronobenzoate, 4-methoxycarbonylbenzeneboronic acid, p-methoxycarbonyl phenylboronic acid, 4-methoxycabonyl phenylboronic acid
PubChem CID 2734369
IUPAC Name (4-methoxycarbonylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)OC)(O)O

Specifications

Melting Point 220°C to 225°C
Quantity 1 g
Solubility Information Soluble in methanol.
Formula Weight 179.97
Percent Purity 97%
Chemical Name or Material 4-(Methoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.