missing translation for 'onlineSavingsMsg'
Learn More

4-(Methoxycarbonyl)phenylboronic Acid, TRC

Product Code. 30467827
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30467827 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30467827 Supplier Toronto Research Chemicals Supplier No. TRCM2616755G
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(Methoxycarbonyl)phenylboronic Acid
CAS 99768-12-4
Molecular Formula C8H9BO4
Quantity 5 g
Synonym (4-Carbomethoxyphenyl)boronic Acid, 4-Carbomethoxybenzeneboronic Acid, 4-Methoxycarbonylbenzeneboronic Acid, Methyl 4-Boronobenzoate, Methyl p-Boronobenzoate, p-(Methoxycarbonyl)boronic Acid, p-(Methoxycarbonyl)phenylboronic Acid
InChI Formula InChI=1S/C8H9BO4/c1-13-8(10)6-2-4-7(5-3-6)9(11)12/h2-5,11-12H,1H3
SMILES COC(=O)c1ccc(cc1)B(O)O
IUPAC Name (4-methoxycarbonylphenyl)boronic acid
Molecular Weight (g/mol) 179.97
Formula Weight 180.0594
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.