missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyindole, 98%, Thermo Scientific™

Product Code. 10705971 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
500mg; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10705971 500mg; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10705971 Supplier Thermo Scientific Acros Supplier No. 133905000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4837-90-5
CAS Min % 97.5
CAS Max % 100.0
Boiling Point 181°C to 183°C (24.0mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NO
MDL Number MFCD00009737
Quantity 500mg
Beilstein 21, II, 44
Synonym 4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe
InChI Key LUNOXNMCFPFPMO-UHFFFAOYSA-N
SMILES COC1=CC=CC2=C1C=CN2
IUPAC Name 4-methoxy-1H-indole
Molecular Weight (g/mol) 147.18
PubChem CID 138363
Formula Weight 147.18
Percent Purity 98%
Chemical Name or Material 4-Methoxyindole, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.