missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyindole, 99%, Thermo Scientific Chemicals

Product Code. 11369615
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11369615 1 g 1g
11389615 250 mg 250mg
11379615 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11369615 Supplier Thermo Scientific Alfa Aesar Supplier No. L19069.03
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4837-90-5
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00009737
InChI Key LUNOXNMCFPFPMO-UHFFFAOYSA-N
Synonym 4-methoxyindole, 1h-indole, 4-methoxy, 4-methoxylindole, 1h-indol-4-yl methyl ether, indole, 4-methoxy, 4-methoxy-indole, zlchem 710, pubchem7433, acmc-1aoxe
PubChem CID 138363
IUPAC Name 4-methoxy-1H-indole
SMILES COC1=CC=CC2=C1C=CN2

Specifications

Melting Point 69°C to 71°C
Boiling Point 181°C to 183°C (24 mmHg)
Quantity 1 g
Beilstein 122456
Sensitivity Light sensitive
Formula Weight 147.18
Percent Purity 99%
Chemical Name or Material 4-Methoxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.