missing translation for 'onlineSavingsMsg'
Learn More

(4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride, 97%, Thermo Scientific™

Product Code. 10263712
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10263712 250 mg 250mg
10160752 1 g 1g
10673445 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10263712 Supplier Thermo Scientific Maybridge Supplier No. SEW03999CB
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 25519-82-8
Molecular Formula C13H18ClNO2
Molecular Weight (g/mol) 255.742
MDL Number MFCD00114817
InChI Key BBDTWYQCXXFKDH-UHFFFAOYSA-N
Synonym 4-4-methoxybenzoyl piperidine hydrochloride, 4-methoxyphenyl 4-piperidyl methanone hydrochloride, 4-methoxyphenyl piperidin-4-yl methanone hydrochloride, methanone, 4-methoxyphenyl-4-piperidinyl-, hydrochloride, 4-4-methoxybenzoyl piperidine hcl, 4-4-methoxybenzoyl-piperidine hydrochloride, 4-methoxy-phenyl-piperidin-4-yl-methanone hydrochloride, ksc801q4b, 4-methoxyphenyl-4-piperidyl methanonehcl, 4-methoxyphenyl 4-piperidyl ketone, chloride
PubChem CID 2740587
IUPAC Name (4-methoxyphenyl)-piperidin-4-ylmethanone;hydrochloride
SMILES COC1=CC=C(C=C1)C(=O)C2CCNCC2.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 255.74
Percent Purity 97%
Chemical Name or Material (4-Methoxyphenyl)(4-piperidyl)methanone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.