missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetone, 98+%, Thermo Scientific™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
122-84-9
C10H12O2
164.2
MFCD00008773
WFWKNGZODAOLEO-UHFFFAOYSA-N
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
31231
1-(4-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=C(C=C1)OC
122-84-9
C10H12O2
164.2
MFCD00008773
WFWKNGZODAOLEO-UHFFFAOYSA-N
4-methoxyphenylacetone, p-acetonylanisole, anisketone, 1-4-methoxyphenyl propan-2-one, 2-propanone, 1-4-methoxyphenyl, anisyl methyl ketone, p-methoxyphenylacetone, 4-methoxybenzyl methyl ketone, 1-p-methoxyphenyl-2-propanone, anisic ketone
31231
1-(4-methoxyphenyl)propan-2-one
CC(=O)CC1=CC=C(C=C1)OC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

CAS Min % 98.0
CAS Max % 100.0
Density 1.0670g/mL
Color Green-Yellow to Yellow
Boiling Point 266°C to 268°C
Flash Point 135°C
Assay Percent Range 98% min. (GC)
Infrared Spectrum Authentic
Linear Formula CH3OC6H4CH2COCH3
Beilstein 08, IV, 460
Packaging Glass bottle
Refractive Index 1.5200 to 1.53
Quantity 250g
Specific Gravity 1.067
Formula Weight 164.2
Physical Form Clear light yellow to yellow-green liquid
Percent Purity 98+%
Chemical Name or Material 4-Methoxyphenylacetone, 98+%
Show More Show Less

RUO – Research Use Only