missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylacetyl chloride, 98%

Product Code. 10714953 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2g
Product Code. Quantity Packaging unitSize
10714953 10 g Glass bottle 10g
10415302 2 g Glass bottle 2g
2 options
This item is not returnable. View return policy

Product Code. 10714953

Brand: Thermo Scientific Acros 368930100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4693-91-8 | C9H9ClO2 | 184.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4693-91-8
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.62
MDL Number MFCD00075474
InChI Key CXJOONIFSVSFAD-UHFFFAOYSA-N
Synonym 4-methoxyphenylacetyl chloride, 2-4-methoxyphenyl acetyl chloride, benzenacetyl chloride, 4-methoxy, benzeneacetyl chloride, 4-methoxy, 4-methoxyphenylacetylchloride, p-methoxyphenyl acetyl chloride, p-methoxyphenylacetyl chloride, acmc-1aso1, 4-methoxyphenylacetylchlorid, 4-methoxybenzeneacetyl chloride
PubChem CID 2734688
IUPAC Name 2-(4-methoxyphenyl)acetyl chloride
SMILES COC1=CC=C(C=C1)CC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.2080g/mL
Boiling Point 143.0°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H4CH2COCl
Refractive Index 1.5390 to 1.5410
Quantity 10 g
Beilstein 10, III, 434
Specific Gravity 1.208
Solubility Information Solubility in water: reacts
Formula Weight 184.62
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methoxyphenylacetyl chloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Methoxyphenylacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.