missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylboronic acid, 97%

Product Code. 10415365 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10415365 5 g Glass bottle 5g
10328790 1 g Glass bottle 1g
10297343 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10415365 Supplier Thermo Scientific Acros Supplier No. 309480050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5720-07-0 | C7H9BO3 | 151.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5720-07-0
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00039139
InChI Key VOAAEKKFGLPLLU-UHFFFAOYSA-N
Synonym 4-methoxyphenyl boronic acid, 4-methoxybenzeneboronic acid, p-anisylboronic acid, p-methoxyphenylboronic acid, p-methoxybenzeneboronic acid, 4-methoxyphenyl boranediol, benzeneboronic acid, p-methoxy, 4-boronoanisole, 4-methoxyphenylboronicacid
PubChem CID 201262
IUPAC Name (4-methoxyphenyl)boronic acid
SMILES COC1=CC=C(C=C1)B(O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 208.0°C to 210.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3OC6H4B(OH)2
Quantity 5 g
Formula Weight 151.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methoxyphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methoxyphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H335-May cause respiratory irritation.
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.