missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxypyridine N-oxide hydrate, 99%, Thermo Scientific™

Product Code. 11311687
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11311687 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11311687 Supplier Thermo Scientific Alfa Aesar Supplier No. L04709.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Methoxypyridine N-oxide hydrate is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207511-18-0
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.142
MDL Number MFCD00149410
InChI Key ABJMARLKAXBQBC-UHFFFAOYSA-N
Synonym 4-methoxypyridine n-oxide hydrate, 4-methoxypyridin-1-ium-1-olate hydrate, 4-methoxypyridine-n-oxide hydrate, c6h7no2.h2o, 4-methoxypyridin-1-ol, hydrate, 4-methoxypyridine-1-oxide hydrate, 4-methoxypyridine-n-oxide hydrate,99, 4-methoxy-1-oxidopyridin-1-ium hydrate, 4-methoxy-1-oxidanidyl-pyridin-1-ium hydrate
PubChem CID 16212293
IUPAC Name 4-methoxy-1-oxidopyridin-1-ium;hydrate
SMILES COC1=CC=[N+](C=C1)[O-].O

Specifications

Melting Point 78°C to 82°C
Quantity 5 g
Beilstein 115212
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 125.13 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 4-Methoxypyridine N-oxide hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.