missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,10-phenanthroline

Product Code. 11378297
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11378297

Brand: Thermo Scientific Alfa Aesar 031888.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31301-28-7 | C13H10N2 | 194.237 g/mol

4-methyl-1,10-phenanthroline can be used to produce 1,10-Phenanthrolin-4-aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31301-28-7
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00004978
InChI Key NAZZKEZTSOOCSZ-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 4-methyl, 4-methylpyridino 3,2-h quinoline, acmc-1aihj, 4-methyl-1,10 phenanthroline, 4-methyl-1,10-phenanthroline, 4-methyl-1,10-phenanthroline1,10-phenanthroline, 4-methyl
PubChem CID 93149
IUPAC Name 4-methyl-1,10-phenanthroline
SMILES CC1=C2C=CC3=C(C2=NC=C1)N=CC=C3

Specifications

Melting Point 144°C to 145°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 194.24
Physical Form Crystalline
Chemical Name or Material 4-Methyl-1,10-phenanthroline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.