missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,10-phenanthroline

Product Code. 11388297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388297 5 g 5g
11378297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388297 Supplier Thermo Scientific Alfa Aesar Supplier No. 031888.06
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31301-28-7 | C13H10N2 | 194.237 g/mol

4-methyl-1,10-phenanthroline can be used to produce 1,10-Phenanthrolin-4-aldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31301-28-7
Molecular Formula C13H10N2
Molecular Weight (g/mol) 194.237
MDL Number MFCD00004978
InChI Key NAZZKEZTSOOCSZ-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 4-methyl, 4-methylpyridino 3,2-h quinoline, acmc-1aihj, 4-methyl-1,10 phenanthroline, 4-methyl-1,10-phenanthroline, 4-methyl-1,10-phenanthroline1,10-phenanthroline, 4-methyl
PubChem CID 93149
IUPAC Name 4-methyl-1,10-phenanthroline
SMILES CC1=C2C=CC3=C(C2=NC=C1)N=CC=C3

Specifications

Melting Point 144°C to 145°C
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 194.24
Physical Form Crystalline
Chemical Name or Material 4-Methyl-1,10-phenanthroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.