missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,3-thiadiazole-5-methanol, 99%, Thermo Scientific™

Product Code. 15493397
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493397 250 mg 250mg
11302129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493397 Supplier Thermo Scientific Alfa Aesar Supplier No. H26129.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos

Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 163008-86-4
Molecular Formula C4H6N2OS
Molecular Weight (g/mol) 130.165
MDL Number MFCD07371384
InChI Key MQLRVKHKJCRSBO-UHFFFAOYSA-N
PubChem CID 18411623
IUPAC Name (4-methylthiadiazol-5-yl)methanol
SMILES CC1=C(SN=N1)CO

Especificaciones

Refractive Index 1.5525
Quantity 250 mg
Synonym 4-methyl-1,2,3-thiadiazol-5-yl methanol, 4-methyl-1,2,3-thiadiazole-5-methanol, 1,2,3-thiadiazole-5-methanol, 4-methyl, 4-methyl-1,2,3-thiadiazol-5-yl methan-1-ol, acmc-209xge, 4-methyl-5-thiadiazolyl methanol, 4-methyl-1,2,3-thiadiazol-5-ylmethanol, 1,2,3-thiadiazole-5-methanol,4-methyl, 5-hydroxymethyl-4-methyl-1,2,3-thiadiazole
Formula Weight 130.17
Percent Purity 99%
Chemical Name or Material 4-Methyl-1,2,3-thiadiazole-5-methanol
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.