missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,2,5-oxadiazole-3-carboxylic Acid, TRC

Product Code. 30477573
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477573 50 mg 50mg
30473005 100 mg 100mg
30482384 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477573 Supplier Toronto Research Chemicals Supplier No. TRCM33614350MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58677-34-2
Molecular Formula C4H4N2O3
Molecular Weight (g/mol) 128.09
Synonym 4-Methyl-1,2,5-oxadiazole-3-carboxylic Acid, 3-Carboxy-4-methylfurazan, 4-Methylfurazan-3-carboxylic Acid, NSC 234497
SMILES Cc1nonc1C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H4N2O3/c1-2-3(4(7)8)6-9-5-2/h1H3,(H,7,8)
Formula Weight 128.0222
Chemical Name or Material 4-Methyl-1,2,5-oxadiazole-3-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.