missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1,3-thiazole-5-sulfonyl chloride, 97%

Product Code. 15284308 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15284308 1 g 1g
15294308 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15284308 Supplier Thermo Scientific Acros Supplier No. 453640010
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 953070-51-4 | C4H4ClNO2S2 | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 953070-51-4
Molecular Formula C4H4ClNO2S2
Molecular Weight (g/mol) 197.66
InChI Key AWTJUYXVSWZCIK-UHFFFAOYSA-N
Synonym 4-methylthiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl, chloro 4-methyl 1,3-thiazol-5-yl sulfone
PubChem CID 18537519
IUPAC Name 4-methyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC=N1)S(=O)(=O)Cl

Specifications

Melting Point 34.0°C to 36.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 197.66
Percent Purity 97%
Chemical Name or Material 4-Methyl-1,3-thiazole-5-sulfonyl chloride
compliance-icons
Product Identifier
  • 4-Methyl-1,3-thiazole-5-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.