missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-cyclohexen-1-ylboronic acid, 97%

Product Code. 11925891 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11925891 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11925891 Supplier Thermo Scientific Acros Supplier No. 431430010
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 850567-92-9 | C7H13BO2 | 139.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 850567-92-9
CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 159.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C7H13BO2
MDL Number MFCD06659857
Quantity 1 g
Synonym 4-methyl-1-cyclohexen-1-ylboronic acid, 4-methylcyclohex-1-en-1-yl boronic acid, 4-methylcyclohex-1-en-1-ylboronic acid, 4-methylcyclohex-1-enylboronic acid, boronic acid, 4-methyl-1-cyclohexen-1-yl, boronic acid,b-4-methyl-1-cyclohexen-1-yl, acmc-209q0u, 4-methyl cyclohexen-1-ylboronic acid, 4-methylcyclohexen-1-yl boronic acid, 4-methyl-1-cyclohexenyl boronic acid
InChI Key SJZMIZIVYIOMIW-UHFFFAOYNA-N
SMILES CC1CCC(=CC1)B(O)O
IUPAC Name (4-methylcyclohexen-1-yl)boronic acid
Molecular Weight (g/mol) 139.99
PubChem CID 44119556
Formula Weight 139.99
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Methyl-1-cyclohexen-1-ylboronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.