missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthaleneboronic acid, 96%, Thermo Scientific Chemicals

Product Code. 11975981 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11975981 1 g 1g
11985981 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Artikelnummer. 11975981 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 437530010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 103986-53-4
Molecular Formula C11H11BO2
Molecular Weight (g/mol) 186.02
MDL Number MFCD01632204
InChI Key JHVQEUGNYSVSDH-UHFFFAOYSA-N
Synonym 4-methyl-1-naphthaleneboronic acid, 4-methyl-1-naphthalene boronic acid, 4-methylnaphthalene-1-boronic acid, 1-methylnaphthalene-4-boronic acid, 4-methylnaphthalen-1-yl boronic acid, 1-borono-4-methylnaphthalene, 4-methyl-1-naphthyl boronic acid, 4-methyl-1-naphthylboronic acid, 4-methylnaphthalen-1-yl boranediol, boronic acid, 4-methyl-1-naphthalenyl
PubChem CID 2773511
IUPAC Name (4-methylnaphthalen-1-yl)boronic acid
SMILES CC1=CC=C(B(O)O)C2=CC=CC=C12

Spezifikation

CAS Min % 95.0
CAS Max % 100.0
Melting Point 224°C to 228°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 186.02
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 4-Methyl-1-naphthaleneboronic acid
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 4-Methyl-1-naphthaleneboronic acid
Signalwort
  • Warning
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.