missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1H-imidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10141132
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. Quantity unitSize
10141132 1 g 1g
10203762 10 g 10g
2 options
This item is not returnable. View return policy

Product Code. 10141132

Brand: Thermo Scientific Maybridge SB01366DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 68282-53-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00173728
InChI Key KMWCSNCNHSEXIF-UHFFFAOYSA-N
Synonym 4-methyl-1h-imidazole-5-carbaldehyde, 5-methylimidazole-4-carboxaldehyde, 4-methylimidazole-5-carboxaldehyde, 4-methyl-5-imidazolecarboxaldehyde, 5-methyl-3h-imidazole-4-carbaldehyde, 4-methylimidazole-5-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 5-methyl, 1h-imidazole-5-carboxaldehyde, 4-methyl, pubchem23016, acmc-1b6zs
PubChem CID 2795887
IUPAC Name 5-methyl-1H-imidazole-4-carbaldehyde
SMILES CC1=C(C=O)N=CN1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 110.12
Percent Purity 97%
Chemical Name or Material 4-Methyl-1H-imidazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.