missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2,1,3-benzoxadiazole, 95%, Thermo Scientific™

Product Code. 11946851 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11946851 5 g 5g
11936851 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11946851 Supplier Thermo Scientific Acros Supplier No. 441360050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 29091-40-5
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD00100600
InChI Key PTXJEYIAUIOTSH-UHFFFAOYSA-N
Synonym 4-methylbenzo c 1,2,5 oxadiazole, 4-methylbenzofurazan, 4-methyl-benzofurazan, 4-methyl-benzo 1,2,5 oxadiazole, 2,1,3-benzoxadiazole, 4-methyl, methylbenzfurazan, 2,1,3-benzoxadiazole,4-methyl, #
PubChem CID 584114
IUPAC Name 4-methyl-2,1,3-benzoxadiazole
SMILES CC1=CC=CC2=NON=C12

Especificaciones

CAS Min % 94.0
CAS Max % 100.0
Melting Point 40.0°C to 41.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 134.14
Percent Purity 95%
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material 4-Methyl-2, 1, 3-benzoxadiazole
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 4-Methyl-2,1,3-benzoxadiazole
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.