missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-[1-(tert-butoxycarbonyl)piperid-4-yl]-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10494163
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10494163 250 mg Amber Glass Bottle 250mg
10269702 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10494163 Supplier Thermo Scientific Maybridge Supplier No. CC31101CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 216955-61-2
Molecular Formula C15H22N2O4S
Molecular Weight (g/mol) 326.41
MDL Number MFCD06659070
InChI Key XDKKXEMKXSGKOR-UHFFFAOYSA-N
Synonym 4-methyl-2-1-tert-butoxycarbonyl piperid-4-yl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxy carbonyl piperidin-4-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-4-methylthiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl-4-piperidinyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-1-tert-butoxycarbonyl piperidin-4-yl-5-carboxy-4-methyl-1,3-thiazole, tert-butyl 4-5-carboxy-4-methyl-1,3-thiazol-2-yl piperidin-1-carboxylate, 4-methyl-2-1-2-methylpropan-2-yl oxycarbonyl piperidin-4-yl-1,3-thiazole-5-carboxylic acid
PubChem CID 2795533
SMILES CC1=C(SC(=N1)C1CCN(CC1)C(=O)OC(C)(C)C)C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name 4-methyl-2-[1-[(2-methylpropan-2-yl)oxycarbonyl]piperidin-4-yl]-1,3-thiazole-5-carboxylic acid
Formula Weight 326.41
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-[1-(tert-butoxycarbonyl)piperid-4-yl]-1, 3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.