missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-(methylthio)pyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 11452150
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452150 5 g 5g
11442150 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11452150 Supplier Thermo Scientific Alfa Aesar Supplier No. H28795.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14001-63-9
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.204
MDL Number MFCD00023242
InChI Key UCERVHYBSTYCQS-UHFFFAOYSA-N
Synonym 4-methyl-2-methylthio pyrimidine, 4-methyl-2 methylsulfanyl pyrimidine, 4-methyl-2-methylsulfanyl pyrimidine, pyrimidine, 4-methyl-2-methylthio, 4-methyl-2-methylsulfanyl-pyrimidine, 2-methylthio-4-methylpyrimidine, 4-methyl-2-methylthiopyrimidine, 4-methyl-2-methylsulphanyl pyrimidine, acmc-1bust, methylmethylsulfanylpyrimidine
PubChem CID 821261
IUPAC Name 4-methyl-2-methylsulfanylpyrimidine
SMILES CC1=NC(=NC=C1)SC

Specifications

Density 1.01
Flash Point 104°C (219°F)
Refractive Index 1.571
Quantity 5 g
Formula Weight 140.21
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-(methylthio)pyrimidine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.