missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-nitroaniline, TRC

Product Code. 30465664
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465664 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465664 Supplier Toronto Research Chemicals Supplier No. TRCM32345050G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Amino-3-nitrotoluene (4-Methyl-2-nitroaniline)
CAS 89-62-3
Molecular Formula C7 H8 N2 O2
Quantity 50 g
Synonym p-Toluidine','2-nitro- (8CI), 4-Methyl-2-nitrobenzenamine, (4-Methyl-2-nitrophenyl)amine, 1-Amino-2-nitro-4-methylbenzene, 2-Nitro-4-methylaniline, 2-Nitro-p-toluidine, 3-Nitro-4-aminotoluene, 4-Amino-3-nitrotoluene, 4-Methyl-2-nitroaniline, 4-Methyl-6-nitroaniline, 4-Methyl-o-nitroaniline, Amarthol Fast Red GL Base, Amarthol Fast Red GL Salt, Azoamine Red A, Azobase NAT, Azobase Red NAT, Azoene Fast Red Red GL Salt, Azofix Red GL Salt, Azoic Diazo Component 8, C.I. 37110, C.I. Azoic Diazo Component 8, Conazoic Diazo M, Devol Red G, Devol Red Salt G, Diazo Fast Red GL, Dycosbase Red GL Base, F. Red Gl Base, Fast Red 3NT Base, Fast Red 3NT Salt, Fast Red Base GL, Fast Red Base JL, Fast Red G Base, Fast Red GL, Fast Red GL Base, Fast Red MGL Base, HD Fast Red GL Base, Hiltonil Fast Red GL Base, Hiltosal Fast Red GL Salt, Lake Red G Base, Lithosol Scarlet Base M, Lithosol Scarlet Base MB, Lithosol Scarlet Base MBW, Lithosol Scarlet Base MW, MNPT, Mitsui Red GL Base, NSC 2759, Naphthanil Red G Base, Naphtoelan Fast Red GL Base, Red Base Ciba VII, Red Base Irga VII, Red Base NGL, Red G Base, Red G Salt, Red Salt Ciba VII, Red Salt Irga VII, Sanyo Fast Red GL Base, Shinnippon Fast Red GL Base, Toyo Fast Red GL Base, Tulabase Fast Red GL, o-Nitro-p-methylaniline, p-Methyl-o-nitroaniline
InChI Formula InChI=1S/C7H8N2O2/c1-5-2-3-6(8)7(4-5)9(10)11/h2-4H,8H2,1H3
SMILES Cc1ccc(N)c(c1)[N+](=O)[O-]
IUPAC Name 4-methyl-2-nitroaniline
Molecular Weight (g/mol) 152.15
Formula Weight 152.0586
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.