missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%, Thermo Scientific Chemicals

Product Code. 10327901 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10327901 10 g Glass bottle 10g
10192811 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10327901 Supplier Thermo Scientific Acros Supplier No. 189710100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332360-07-3 , 4502-00-5
Molecular Formula C6H9NaO3·xH2O
Molecular Weight (g/mol) 152.13
MDL Number MFCD00150776
InChI Key FUDIEMDWYRETTC-UHFFFAOYSA-M
Synonym 4-methyl-2-oxopentanoic acid sodium salt hydrate, sodium hydrate ?-ketoisocaproate, 4-methyl-2-oxopentanoic acid,sodium salt,hydrate,98+%
PubChem CID 23680490
IUPAC Name sodium;4-methyl-2-oxopentanoate;hydrate
SMILES CC(C)CC(=O)C(=O)[O-].O.[Na+]

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 280°C
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (CH3)2CHCH2COCO2Na·xH2O
Quantity 10 g
Solubility Information Solubility in water: soluble
Formula Weight 152.13
Percent Purity 98+%
Chemical Name or Material 4-Methyl-2-oxopentanoic acid, sodium salt, hydrate, 98+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.