missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-oxovaleric acid, 94%

Product Code. 15496538
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15496538 5 g 5g
15486538 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496538 Supplier Thermo Scientific Alfa Aesar Supplier No. H60076.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 816-66-0 | C6H9NaO3 | 152.13 g/mol

Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ii is an intermediate in the metabolism of leucine and also used as flavoring agents.

Solubility
Soluble in water.

Notes
Refrigerate in tightly sealed containers. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 816-66-0
Molecular Formula C6H9NaO3
Molecular Weight (g/mol) 152.13
MDL Number MFCD00066204
InChI Key IXFAZKRLPPMQEO-UHFFFAOYSA-M
Synonym 4-methyl-2-oxovaleric acid, alpha-ketoisocaproic acid, ketoleucine, alpha-ketoisocaproate, pentanoic acid, 4-methyl-2-oxo, 2-oxoisocaproate, 2-oxo-4-methylpentanoic acid, ketoisocaproate, 2-oxoisohexanoate, 4-methyl-2-oxopentanoate
PubChem CID 70
ChEBI CHEBI:48430
SMILES [Na+].CC(C)CC(=O)C([O-])=O

Specifications

Melting Point 8°C to 10°C
Density 1.055
Boiling Point 82°C to 83°C
Linear Formula (CH3)2CHCH2COCOOH
Refractive Index 1.431
Quantity 5 g
UN Number UN3265
Beilstein 1701823
Solubility Information Soluble in water.
IUPAC Name 4-methyl-2-oxopentanoic acid
Formula Weight 130.14
Percent Purity 98%
Chemical Name or Material 4-Methyl-2-oxovaleric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.