missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-oxovaleric Acid Sodium Salt, TRC

Product Code. 30133073
Change view
Click to view available options
Quantity:
500 mg
1 g
25 g
Unit Size:
1g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133073 500 mg 500mg
30132970 25 g 25g
30007525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133073 Supplier Toronto Research Chemicals Supplier No. TRCM326425500MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4502-00-5
Molecular Formula C6 H9 O3 . Na
Molecular Weight (g/mol) 152.12
Synonym Pentanoic acid, 4-methyl-2-oxo-, sodium salt (1:1), Pentanoic acid, 4-methyl-2-oxo-, sodium salt (9CI), Valeric acid, 4-methyl-2-oxo-, sodium salt (8CI), 2-Ketoisocaproic acid sodium salt, 2-Oxo-4-methylpentanoic acid sodium salt, 4-Methyl-2-oxopentanoic acid sodium salt, Sodium 2-ketoisocaproate, Sodium 4-methyl-2-oxopentanoate, Sodium 4-methyl-2-oxovalerate, Sodium α-keto-isocaproate, Sodium α-ketoisocaproate, Sodium α-oxoisocaproate
IUPAC Name sodium;4-methyl-2-oxopentanoate
SMILES [Na+].CC(C)CC(=O)C(=O)[O-]

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H10O3.Na/c1-4(2)3-5(7)6(8)9;/h4H,3H2,1-2H3,(H,8,9);/q;+1/p-1
Formula Weight 152.0449
Chemical Name or Material 4-Methyl-2-oxopentanoic acid sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.