missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanol, TRC

Product Code. 17930935
Change view
Click to view available options
Quantity:
50 mL
500 mL
Unit Size:
500mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17930935 50 mL 50mL
17922135 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17930935 Supplier Toronto Research Chemicals Supplier No. TRCM32475050ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-11-2
Molecular Formula C6 H14 O
Molecular Weight (g/mol) 102.17
Synonym 4-Methyl-pentan-2-ol, 4-Methylpentan-2-ol, 2-Methyl-4-pentanol, (±)-4-Methyl-2-pentanol, 1,3-Dimethyl-1-butanol, 1,3-Dimethylbutyl alcohol, 2-Methyl-4-pentanol, 3-MIC, 4-Methyl-2-amyl alcohol, 4-Methyl-2-pentyl alcohol, Isobutylmethylcarbinol, Isobutylmethylmethanol, MAOH, MIBC, MIC, Methylisobutylcarbinol, NSC 9384, dl-Methylisobutylcarbinol
IUPAC Name 4-methylpentan-2-ol
SMILES CC(C)CC(C)O

Specifications

Quantity 50 mL
InChI Formula InChI=1S/C6H14O/c1-5(2)4-6(3)7/h5-7H,4H2,1-3H3
Formula Weight 102.1045
Chemical Name or Material 2-Methyl-4-pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.