missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone-d5, TRC

Product Code. 30478805
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30478805 1 g 1g
30466307 100 mg 100mg
30474814 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30478805 Supplier Toronto Research Chemicals Supplier No. TRCM3280271G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4840-81-7
Molecular Formula C6 2H5 H7 O
Molecular Weight (g/mol) 105.19
Synonym 4-Methyl-2-pentanone D5 (1,1,1,3,3-D5), 2-Pentanone-1,1,1,3,3-d5, 4-methyl- (7CI, 8CI, 9CI)
IUPAC Name 1,1,1,3,3-pentadeuterio-4-methylpentan-2-one
SMILES [2H]C([2H])([2H])C(=O)C([2H])([2H])C(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3/i3D3,4D2
Formula Weight 105.1202
Chemical Name or Material 4-Methyl-2-pentanone-d5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.