missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-pentanone, TRC

Product Code. 30483194
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30483194 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30483194 Supplier Toronto Research Chemicals Supplier No. TRCM328025100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methyl-2-pentanone (MIBK)
CAS 108-10-1
Purity Grade Notes GC
Molecular Formula C6 H12 O
Quantity 100 g
Synonym 2-Pentanone, 4-methyl-, 4-Methyl-2-pentanone, 2-Methyl-4-pentanone, 2-Methylpropyl methyl ketone, 4-Methyl-2-oxopentane, Hexone, Isobutyl methyl ketone, Isopropylacetone, MIBK, MIK, Methyl 2-methylpropyl ketone, Methyl iso-butyl ketone, Methyl isobutyl ketone, NSC 5712, ZMD-D
InChI Formula InChI=1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3
SMILES CC(C)CC(=O)C
IUPAC Name 4-methylpentan-2-one
Molecular Weight (g/mol) 100.16
Formula Weight 100.0888
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.