missing translation for 'onlineSavingsMsg'
Learn More

(4-Methyl-2-phenyl-1,3-thiazol-5-yl)methanol, 97%, Thermo Scientific™

Product Code. 10036242
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10036242 10 g Amber Glass Bottle 10g
10710354 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10036242 Supplier Thermo Scientific Maybridge Supplier No. CC29209EA
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 61291-91-6
Molecular Formula C11H11NOS
Molecular Weight (g/mol) 205.275
MDL Number MFCD01928782
InChI Key JSIPFWSPCWZJIU-UHFFFAOYSA-N
Synonym 4-methyl-2-phenyl-1,3-thiazol-5-yl methanol, 4-methyl-2-phenylthiazol-5-yl methanol, 4-methyl-2-phenyl-thiazol-5-yl-methanol, 4-methyl-2-phenyl-1,3-thiazol-5-yl methan-1-ol, 4-methyl-2-phenyl-5-thiazolemethanol, 5-hydroxymethyl-4-methyl-2-phenylthiazole
PubChem CID 736541
IUPAC Name (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.28
Percent Purity 97%
Chemical Name or Material (4-Methyl-2-phenyl-1, 3-thiazol-5-yl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.