missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-carbaldehyde, Thermo Scientific™

Product Code. 10473403
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10473403 250 mg 250mg
10025762 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10473403 Supplier Thermo Scientific Maybridge Supplier No. CC29204CB
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 55327-23-6
Molecular Formula C11H9NOS
Molecular Weight (g/mol) 203.26
MDL Number MFCD04974047
InChI Key QRPSQJOXOZJEHJ-UHFFFAOYSA-N
Synonym 4-methyl-2-phenylthiazole-5-carbaldehyde, 4-methyl-2-phenyl-thiazole-5-carbaldehyde, 2-phenyl-4-methylthiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde, 4-methyl-2-phenyl
PubChem CID 2795491
IUPAC Name 4-methyl-2-phenyl-1,3-thiazole-5-carbaldehyde
SMILES CC1=C(SC(=N1)C1=CC=CC=C1)C=O

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 203.26
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.