missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride, Technical, Thermo Scientific™

Product Code. 10337734
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10337734 5 g Amber Glass Bottle 5g
10641275 250 mg Amber Glass Bottle 250mg
10041222 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10337734 Supplier Thermo Scientific Maybridge Supplier No. CC29203DE
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-88-3
Molecular Formula C10H8ClNO2S2
Molecular Weight (g/mol) 273.749
InChI Key NGDQQLAVJWUYSF-UHFFFAOYSA-N
Synonym 4-methyl-2-phenylthiazole-5-sulfonyl chloride, 4-methyl-2-phenyl-thiazole-5-sulfonyl chloride, 5-thiazolesulfonyl chloride, 4-methyl-2-phenyl, 4-methyl-2-phenyl-1,3-thiazole-5-sulfonylchloride
PubChem CID 2795490
IUPAC Name 4-methyl-2-phenyl-1,3-thiazole-5-sulfonyl chloride
SMILES CC1=C(SC(=N1)C2=CC=CC=C2)S(=O)(=O)Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 273.76
Percent Purity 95%
Chemical Name or Material 4-Methyl-2-phenyl-1, 3-thiazole-5-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.