missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Product Code. 11300587
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11300587 5 g 5g
10782663 1 g 1g
10564505 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11300587 Supplier Thermo Scientific Maybridge Supplier No. CC20201DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 103249-79-2
Molecular Formula C12H10N2O2
Molecular Weight (g/mol) 214.224
InChI Key ZPXPNKCBUXSJFK-UHFFFAOYSA-N
Synonym 4-methyl-2-phenyl-5-pyrimidinecarboxylic acid, 5-pyrimidinecarboxylicacid, 4-methyl-2-phenyl, acmc-1c8q0, 2-phenyl-4-methylpyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid,4-methyl-2-phenyl, 4-methyl-2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylic acid, 4-methyl-2-phenyl
PubChem CID 239975
IUPAC Name 4-methyl-2-phenylpyrimidine-5-carboxylic acid
SMILES CC1=NC(=NC=C1C(=O)O)C2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 214.22
Percent Purity 97%
Chemical Name or Material 4-Methyl-2-phenyl-5-pyrimidinecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.