missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-[[1-oxo-2-(propylamino)propyl]amino]-2-thiophenecarboxylic Acid-d7, TRC

Product Code. 30477960
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477960 1 mg 1mg
30471938 10 mg 10mg
30471949 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477960 Supplier Toronto Research Chemicals Supplier No. TRCM3268161MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12H11D7N2O3S
Molecular Weight (g/mol) 277.39
Synonym Articiaine Acid-d7, Articainic Acid-d7
SMILES O=C(C(C)NC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])NC1=C(C(O)=O)SC=C1C

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H18N2O3S/c1-4-5-13-8(3)11(15)14-9-7(2)6-18-10(9)12(16)17/h6,8,13H,4-5H2,1-3H3,(H,14,15)(H,16,17)/i1D3,4D2,5D2
Chemical Name or Material 4-Methyl-3-[(1-oxo-2-(propylamino)propyl)amino]-2-thiophenecarboxylic Acid-d7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.