missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10537274
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10537274 250 mg Amber Glass Bottle 250mg
10693615 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10537274 Supplier Thermo Scientific Maybridge Supplier No. CC29801CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 212578-38-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD11506351
InChI Key SSVIRLKDUUXSTR-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
PubChem CID 10845449
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
SMILES CN1CC(OC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.