missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-1,4-benzoxazine-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10693615
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10693615 1 g Amber Glass Bottle 1g
10537274 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10693615 Supplier Thermo Scientific Maybridge Supplier No. CC29801DA
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 212578-38-6
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD11506351
InChI Key SSVIRLKDUUXSTR-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazine-2-carboxylic acid, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazine-2-carboxylic acid, 2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl, 4-methyl-2h,3h-benzo e 1,4-oxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazine-2-carboxylic acid, 3,4-dihydro-4-methyl-2h-1,4-benzoxazole-2-carboxylic acid
PubChem CID 10845449
IUPAC Name 4-methyl-2,3-dihydro-1,4-benzoxazine-2-carboxylic acid
SMILES CN1CC(OC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 193.2
Percent Purity 97%
Chemical Name or Material 4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazine-2-carboxylic acid
Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.