missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10544974
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10544974 250 mg Amber Glass Bottle 250mg
10011434 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10544974 Supplier Thermo Scientific Maybridge Supplier No. CC62001CB
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Identificatori chimici

CAS 915707-58-3
Molecular Formula C9H10N2O3
Molecular Weight (g/mol) 194.19
MDL Number MFCD09065017
InChI Key JTDUYLQIBHWRRK-UHFFFAOYSA-N
Synonym 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 4-methyl-2h,3h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid, 2h-pyrido 3,2-b-1,4-oxazine-7-carboxylicacid, 3,4-dihydro-4-methyl, 4-methyl-3,4-dihydro-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylicacid, 4-methyl-2h,3h-pyridino 2,3-e 1,4-oxazine-7-carboxylic acid, 7-carboxy-3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b-1,4-oxazine-7-carboxylic acid, 3,4-dihydro-4-methyl-2h-pyrido 3,2-b 1,4 oxazine-7-carboxylic acid
PubChem CID 24229645
IUPAC Name 4-methyl-2,3-dihydropyrido[3,2-b][1,4]oxazine-7-carboxylic acid
SMILES CN1CCOC2=C1N=CC(=C2)C(=O)O

Specifica

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 194.19
Percent Purity 97%
Chemical Name or Material 4-Methyl-3,4-dihydro-2H-pyrido[3,2-b][1,4]oxazine-7-carboxylic acid
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.