missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-nitroimidazole, 98%, Thermo Scientific™

Product Code. 11986271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11986271 1 g Glass bottle 1g
11996271 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11986271 Supplier Thermo Scientific Acros Supplier No. 440400010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 14003-66-8
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.10
MDL Number MFCD01721395
InChI Key WSYOWIMKNNMEMZ-UHFFFAOYSA-N
Synonym 4-methyl-5-nitroimidazole, 5-methyl-4-nitroimidazole, 4-methyl-5-nitro-1h-imidazole, 1h-imidazole, 4-methyl-5-nitro, imidazole, 4-methyl-5-nitro, imidazole, 5-methyl-4-nitro, unii-5pul8bw840, imidazole, 4 or 5-methyl-5 or 4-nitro, 4-methyl-5-nitro-3h-imidazole, 1h-imidazole,4-methyl-5-nitro
PubChem CID 26385
IUPAC Name 5-methyl-4-nitro-1H-imidazole
SMILES CC1=C(N=CN1)[N+]([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 203.2°C to 203.3°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 127.1
Percent Purity 98%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 4-Methyl-5-nitroimidazole
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methyl-4-nitroimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.