missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-thiazoleethanol, 98%

Product Code. 10531251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10531251 25 mL Glass bottle 25mL
10092260 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10531251 Supplier Thermo Scientific Acros Supplier No. 184060250
 Restricted Item
Estimated Shipment: 28-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 137-00-8 | C6H9NOS | 143.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-00-8
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.21
MDL Number MFCD00005339
InChI Key BKAWJIRCKVUVED-UHFFFAOYSA-N
Synonym 4-methyl-5-thiazoleethanol, sulfurol, 5-2-hydroxyethyl-4-methylthiazole, hemineurine, 2-4-methylthiazol-5-yl ethanol, 4-methyl-5-thiazolethanol, 5-thiazoleethanol, 4-methyl, 2-4-methyl-1,3-thiazol-5-yl ethanol, 4-methyl-5-hydroxyethylthiazole, 2-4-methyl-1,3-thiazol-5-yl ethan-1-ol
PubChem CID 1136
ChEBI CHEBI:17957
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
SMILES CC1=C(SC=N1)CCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Brown
Density 1.1900g/mL
Boiling Point 135°C (7.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5480 to 1.551
Quantity 25 mL
Beilstein 27, 1754
Merck Index 15, 6199
Specific Gravity 1.19
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol,ether,benzene and chloroform
Formula Weight 143.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Methyl-5-thiazoleethanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Methyl-5-thiazoleethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.