missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-thiazoleethanol, TRC

Product Code. 30004430
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004430 5 g 5g
30011832 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004430 Supplier Toronto Research Chemicals Supplier No. TRCM3302355G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-00-8
Molecular Formula C6 H9 N O S
Molecular Weight (g/mol) 143.21
Synonym 5-(2-Hydroxyethyl)-4-methylthiazole, MHT, 5-Thiazoleethanol, 4-methyl- (6CI, 8CI, 9CI, ACI), 4-Methyl-5-thiazoleethanol (ACI), 2-(4-Methyl-1,3-thiazol-5-yl)ethan-1-ol, 2-(4-Methyl-1,3-thiazol-5-yl)ethanol, 2-(4-Methyl-5-thiazolyl)ethanol, 2-(4-Methylthiazole-5-yl)ethanol, 4-Methyl-5-(2-hydroxyethyl)thiazole, 4-Methyl-5-(2-thiazoleethanol), 4-Methyl-5-(β-hydroxyethyl)thiazole, 4-Methyl-5-thiazolylethanol, 5-(Hydroxyethyl)-4-methylthiazole, 5-(β-Hydroxyethyl)-4-methylthiazole, Hemineurine, NSC 23262, NSC 41831, Sulfurol
IUPAC Name 2-(4-methyl-1,3-thiazol-5-yl)ethanol
SMILES Cc1ncsc1CCO

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H9NOS/c1-5-6(2-3-8)9-4-7-5/h4,8H,2-3H2,1H3
Formula Weight 143.0405
Chemical Name or Material 5-(2-Hydroxyethyl)-4-methylthiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.