missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-5-vinylthiazole, 98+%

Product Code. 11430327
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430327 5 g 5g
11440327 25 g 25g
11450327 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11430327 Supplier Thermo Scientific Alfa Aesar Supplier No. B21719.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1759-28-0 | C6H7NS | 125.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1759-28-0
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00005337
InChI Key QUAMMXIRDIIGDJ-UHFFFAOYSA-N
Synonym 4-methyl-5-vinylthiazole, thiazole, 5-ethenyl-4-methyl, 4-methyl-5-vinyl thiazole, 5-ethenyl-4-methylthiazole, thiazole, 4-methyl-5-vinyl, vinylsulfurol, vinyl sulfurol, fema no. 3313, 5-vinyl-4-methylthiazole, 4-methyl-5-vinyl-thiazole
PubChem CID 15654
IUPAC Name 5-ethenyl-4-methyl-1,3-thiazole
SMILES CC1=C(SC=N1)C=C

Specifications

Melting Point -15°C
Density 1.095
Boiling Point 67°C to 68°C (11 mmHg)
Flash Point 70°C (158°F)
Odor Pleasant
Refractive Index 1.568
Quantity 5 g
Beilstein 107867
Formula Weight 125.19
Percent Purity ≥98%
Chemical Name or Material 4-Methyl-5-vinylthiazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.