missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific™

Product Code. 11346825
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11346825 5 g Amber Glass Bottle 5g
10289232 250 mg Amber Glass Bottle 250mg
10548053 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11346825 Supplier Thermo Scientific Maybridge Supplier No. CC13539DE
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 519054-54-7
Molecular Formula C15H22BNO3
Molecular Weight (g/mol) 275.16
MDL Number MFCD04115377
InChI Key QRAOZQGIUIDZQZ-UHFFFAOYSA-N
Synonym 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine, 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-7-ylboronic acid pinacol ester, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydro-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-dioxaborolan, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-3,4-dihydro-2h-benzo 1,4 oxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2,-dioxaborolan-2-yl-3,4-dihydro-2h-1,4-benzoxazine, 4-methyl-7-4,4,5,5-tetramethyl-1,3,2-d ioxaborolan-2-yl-3,4-dihydro-2h-benzo b 1,4 oxazine
PubChem CID 2795301
IUPAC Name 4-methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-benzoxazine
SMILES CN1CCOC2=C1C=CC(=C2)B1OC(C)(C)C(C)(C)O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 275.15
Chemical Name or Material 4-Methyl-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-1,4-benzoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.