missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl Hippuric Acid, TRC

Product Code. 30478273
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478273 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30478273 Supplier Toronto Research Chemicals Supplier No. TRCM3119755G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methyl Hippuric Acid
CAS 27115-50-0
Purity Grade Notes HPLC
Molecular Formula C10 H11 N O3
Quantity 5 g
Synonym Glycine','N-(4-methylbenzoyl)-, Hippuric acid','p-methyl- (6CI,8CI), N-(4-Methylbenzoyl)glycine, (4-Methylbenzoylamino)acetic acid, 2-[(4-Methylbenzoyl)amino]acetic acid, 2-[(4-Methylphenyl)formamido]acetic acid, 4-Methylhippuric acid, N-(p-Methylbenzoyl)glycine, NSC 126814, [[(4-Methylphenyl)carbonyl]amino]acetic acid, p-Methylhippuric acid, p-Toluric acid, 4-Methyl hippuric acid
InChI Formula InChI=1S/C10H11NO3/c1-7-2-4-8(5-3-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)
SMILES Cc1ccc(cc1)C(=O)NCC(=O)O
IUPAC Name 2-[(4-methylbenzoyl)amino]acetic acid
Molecular Weight (g/mol) 193.2
Formula Weight 193.0739
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.