missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl Phencyclidine Hydrochloride, TRC

Product Code. 30478074
Change view
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30478074 25 mg 25mg
30470484 5 mg 5mg
30477178 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30478074 Supplier Toronto Research Chemicals Supplier No. TRCM32556025MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1934-42-5
Molecular Formula C18 H27 N . Cl H
Molecular Weight (g/mol) 293.87
Synonym 4-Me PCP, 4-Me Phencyclidine, 4-methyl Phencyclidine
IUPAC Name 1-[1-(p-tolyl)cyclohexyl]piperidine;hydrochloride
SMILES Cl.Cc1ccc(cc1)C2(CCCCC2)N3CCCCC3

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H27N.ClH/c1-16-8-10-17(11-9-16)18(12-4-2-5-13-18)19-14-6-3-7-15-19;/h8-11H,2-7,12-15H2,1H3;1H
Formula Weight 293.191
Chemical Name or Material 4-methyl PCP (hydrochloride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.