missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 95%

Product Code. 10636902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10636902 250 g Glass bottle 250g
10215320 5 g Glass bottle 5g
10417710 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10636902 Supplier Thermo Scientific Acros Supplier No. 126182500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-00-9 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -19.0°C
Color Colorless to Yellow
Density 1.0050g/mL
Boiling Point 226.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4COCH3
Refractive Index 1.5320 to 1.5350
Quantity 250 g
Beilstein 07, 307
Specific Gravity 1.005
Solubility Information Solubility in water: 0.37g/L (15°C)
Formula Weight 134.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 4'-Methylacetophenone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Methylacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.