missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 96%

Product Code. 11471788
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471788 50 g 50g
11481788 250 g 250g
11461788 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471788 Supplier Thermo Scientific Alfa Aesar Supplier No. A14469.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122-00-9 | C9H10O | 134.18 g/mol

4'-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1

Spécification

Melting Point -19°C
Density 1.01
Boiling Point 224°C to 226°C
Flash Point 92°C (197°F)
Refractive Index 1.534
Quantity 50 g
Beilstein 606053
Solubility Information Miscible with water.
Formula Weight 134.18
Percent Purity 96%
Chemical Name or Material 4'-Methylacetophenone
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 4'-Methylacetophenone
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Mention de danger
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Informations supplémentaires
  • MIXTURE LIST-Contain: p-Methylacetophenone

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.