missing translation for 'onlineSavingsMsg'
Learn More

4'-Methylacetophenone, 96%

Product Code. 11481788
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481788 250 g 250g
11471788 50 g 50g
11461788 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11481788 Supplier Thermo Scientific Alfa Aesar Supplier No. A14469.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 122-00-9 | C9H10O | 134.18 g/mol

4'-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Methylacetophenone is used as a flavoring agent. It reacts with morpholine to get 4-(p-tolyl-thioacetyl)-morpholine in the presence of sulfur as a reagent. Further, it is used as an intermediate in the manufacture of active pharmaceutical ingredients, perfumes and cosmetics.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-00-9
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00008751
InChI Key GNKZMNRKLCTJAY-UHFFFAOYSA-N
Synonym 4'-methylacetophenone, p-methylacetophenone, 1-p-tolyl ethanone, 1-p-tolylethanone, melilotal, 4-methylacetophenone, 4-acetyltoluene, 1-4-methylphenyl ethanone, methyl p-tolyl ketone, p-acetotoluene
PubChem CID 8500
IUPAC Name 1-(4-methylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C)C=C1

Specifications

Melting Point -19°C
Density 1.01
Boiling Point 224°C to 226°C
Flash Point 92°C (197°F)
Refractive Index 1.534
Quantity 250 g
Beilstein 606053
Solubility Information Miscible with water.
Formula Weight 134.18
Percent Purity 96%
Chemical Name or Material 4'-Methylacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.